Sodium carbonate, 99.6%, ACS reagent, anhydrous
Sodium carbonate, 99.6%, ACS reagent, anhydrous
Sodium carbonate, 99.6%, ACS reagent, anhydrous
Sodium carbonate, 99.6%, ACS reagent, anhydrous
Thermo Scientific Chemicals
Sodium carbonate, 99.6%, ACS reagent, anhydrous
CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA424285000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium carbonate
Name NoteAnhydrous
CAS497-19-8
ColorWhite
Health Hazard 1Warning
View more
Adsorbent
RUO – Research Use Only