Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous
Thermo Scientific Chemicals
Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous
CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number FSA424255000
Price (MYR)
540.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
540.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcetic acid, sodium salt
pH7.0 to 9.2 (5% soln. at 25°C)
CAS127-09-3
CAS Min %99.0
Health Hazard 1HYGROSCOPIC