Potassium nitrate, 99+%, ACS reagent
Potassium nitrate, 99+%, ACS reagent
Potassium nitrate, 99+%, ACS reagent
Potassium nitrate, 99+%, ACS reagent
Thermo Scientific Chemicals
Potassium nitrate, 99+%, ACS reagent
CAS: 7757-79-1 | KNO3 | 101.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA424155000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point400.0°C
Chemical Name or MaterialPotassium nitrate
Melting Point334.0°C
Name Notecrystals
CAS7757-79-1