Phosphoric acid, ACS reagent, 85+% solution in water
Phosphoric acid, ACS reagent, 85+% solution in water
Phosphoric acid, ACS reagent, 85+% solution in water
Phosphoric acid, ACS reagent, 85+% solution in water
Thermo Scientific Chemicals
Phosphoric acid, ACS reagent, 85+% solution in water
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA424045000
Price (MYR)
562.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
562.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhosphoric acid
Name Note85+% Solution in Water
CAS7664-38-2
Melting Point21.0°C
Boiling Point158.0°C