Perchloric acid, ACS reagent, ca. 70% solution in water
Perchloric acid, ACS reagent, ca. 70% solution in water
Perchloric acid, ACS reagent, ca. 70% solution in water
Perchloric acid, ACS reagent, ca. 70% solution in water
Thermo Scientific Chemicals
Perchloric acid, ACS reagent, ca. 70% solution in water
CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA424032500
Price (MYR)
2,036.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,036.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPerchloric acid
Name Noteca. 70% Solution in Water
CAS7601-90-3
Melting Point-18.0°C
Boiling Point203.0°C
View more
Oxidation reagent
RUO – Research Use Only