Nitromethane, 96%, ACS reagent
Nitromethane, 96%, ACS reagent
Nitromethane, 96%, ACS reagent
Nitromethane, 96%, ACS reagent
Thermo Scientific Chemicals
Nitromethane, 96%, ACS reagent
CAS: 75-52-5 | CH3NO2 | 61.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA424015000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNitromethane
Name Note95%
CAS75-52-5
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning