4-Methyl-2-pentanone, 98.5+%, ACS reagent
4-Methyl-2-pentanone, 98.5+%, ACS reagent
4-Methyl-2-pentanone, 98.5+%, ACS reagent
4-Methyl-2-pentanone, 98.5+%, ACS reagent
Thermo Scientific Chemicals
4-Methyl-2-pentanone, 98.5+%, ACS reagent
CAS: 108-10-1 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
1 L
Packaging:
View all
Glass bottle
Catalog number FSA423960025
Price (MYR)
719.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
719.00
EA
Add to cart
Specifications
Chemical Name or Material4-Methyl-2-pentanone
Name NoteFor analysis ACS
CAS108-10-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.