Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Thermo Scientific Chemicals
Methanol, >=99.8%, ACS reagent, meets the requirements of Reag.Ph.Eur.
CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA423955000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethanol
Melting Point-98.0°C
CAS67-56-1
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.