Hydrobromic acid, ACS reagent, ca. 48% solution in water
Hydrobromic acid, ACS reagent, ca. 48% solution in water
Hydrobromic acid, ACS reagent, ca. 48% solution in water
Hydrobromic acid, ACS reagent, ca. 48% solution in water
Thermo Scientific Chemicals
Hydrobromic acid, ACS reagent, ca. 48% solution in water
Hydrobromic acid, 47.0 to 49.0%, BrH, CAS Number-10035-10-6, 7732-18-5, acide bromhydrique, acido bromidrico, anhydrous hydrobromic acid, bromowodor, bromowodor polish, bromwasserstoff, broomwaterstof, hydrobromic acid, hydrobromide, hydrogen bromide, 2.5L, 126.0 deg.C to 128.0 deg.C, CHEBI:47266 | CAS: 10035-10-6 | BrH | 80.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA423785000
Price (MYR)
578.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
578.00
BOT
Add to cart
Specifications
Chemical Name or MaterialHydrobromic acid
ColorColorless to Yellow
CAS10035-10-6
Melting Point-11.0°C
Boiling Point126.0°C to 128.0°C
View more
C-X bond formation reagent
RUO – Research Use Only