Formic acid, 97%, ACS reagent
Formic acid, 97%, ACS reagent
Formic acid, 97%, ACS reagent
Formic acid, 97%, ACS reagent
Thermo Scientific Chemicals
Formic acid, 97%, ACS reagent
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA423755000
Price (MYR)
479.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
479.00
EA
Add to cart
Specifications
Chemical Name or MaterialFormic acid
Name Note96+%
CAS64-18-6
Melting Point8.0°C
Boiling Point101.0°C