4-Dimethylaminobenzaldehyde, ACS reagent
4-Dimethylaminobenzaldehyde, ACS reagent
4-Dimethylaminobenzaldehyde, ACS reagent
4-Dimethylaminobenzaldehyde, ACS reagent
Thermo Scientific Chemicals
4-Dimethylaminobenzaldehyde, ACS reagent
CAS: 100-10-7 | C9H11NO | 149.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA423635000
Price (MYR)
2,029.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,029.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Dimethylaminobenzaldehyde
CAS100-10-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful to aquatic life with long lasting effects. Harmful if swallowed.
Health Hazard 3GHS P Statement
Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.