Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent
Thermo Scientific Chemicals
Cobalt(II) chloride hexahydrate, 98-102%, ACS reagent
CAS: 7791-13-1 | Cl2CoH12O6 | 237.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
View all
Plastic Bottle
Catalog number FSA423575000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt(II) chloride hexahydrate
CAS7791-13-1
ColorLilac
Health Hazard 1Signal Word: DANGER!
Health Hazard 2GHS P Statement May damage fertility. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Suspected of causing genetic defects. May cause cancer by inhalation. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled.
View more
Suitable for battery materials development.
RUO – Research Use Only