Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Citric acid, ≥99.5%, ACS reagent, anhydrous
Thermo Scientific Chemicals
Citric acid, ≥99.5%, ACS reagent, anhydrous
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number FSA423565000
Price (MYR)
592.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
592.00
BOT
Add to cart
Specifications
Chemical Name or MaterialCitric acid
CAS77-92-9
Melting Point153.0°C
Molecular FormulaC6H8O7
Linear FormulaHO2CCH2C(OH)(CO2H)CH2CO2H