1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur.
1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur.
1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur.
1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur.
Thermo Scientific Chemicals
1-Butanol, 99.5%, ACS reagent, meets the requirements of Reag.Ph.Eur.
CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
EcoSafPak™ Glass Bottle
Catalog number FSA423495000
Price (MYR)
383.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
383.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Butanol
CAS71-36-3
CAS Min %99.4
Health Hazard 1Danger
CAS Max %100.0