Benzoic acid, 99.6%, ACS reagent
Benzoic acid, 99.6%, ACS reagent
Benzoic acid, 99.6%, ACS reagent
Benzoic acid, 99.6%, ACS reagent
Thermo Scientific Chemicals
Benzoic acid, 99.6%, ACS reagent
CAS: 65-85-0 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Plastic bottle
Catalog number FSA423475000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialBenzoic acid
CAS65-85-0
CAS Min %99.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0