Ammonium oxalate monohydrate, 99+%, ACS reagent
Ammonium oxalate monohydrate, 99+%, ACS reagent
Ammonium oxalate monohydrate, 99+%, ACS reagent
Ammonium oxalate monohydrate, 99+%, ACS reagent
Thermo Scientific Chemicals
Ammonium oxalate monohydrate, 99+%, ACS reagent
CAS: 6009-70-7 | C2H8N2O4·H2O | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number FSA423365000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium oxalate monohydrate
Melting Point70.0°C
CAS6009-70-7
ColorWhite
Health Hazard 1GHS Signal Word: Warning