Ammonium chloride, 99.6%, ACS reagent
Ammonium chloride, 99.6%, ACS reagent
Ammonium chloride, 99.6%, ACS reagent
Ammonium chloride, 99.6%, ACS reagent
Ammonium chloride, 99.6%, ACS reagent
Thermo Scientific Chemicals
Ammonium chloride, 99.6%, ACS reagent
CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number FSA423285000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium chloride
Melting Point340.0°C
CAS12125-02-9
ColorWhite
Health Hazard 1GHS Signal Word: Warning
View more
Suitable for battery materials development.
RUO – Research Use Only