Oxalic acid dihydrate, 99.5+%, ACS reagent
Oxalic acid dihydrate, 99.5+%, ACS reagent
Oxalic acid dihydrate, 99.5+%, ACS reagent
Oxalic acid dihydrate, 99.5+%, ACS reagent
Thermo Scientific Chemicals
Oxalic acid dihydrate, 99.5+%, ACS reagent
CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number FSA423152500
Price (MYR)
360.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
360.00
EA
Add to cart
Specifications
Chemical Name or MaterialOxalic acid dihydrate
Name NoteFor analysis ACS
CAS6153-56-6
Melting Point98.0°C to 102.0°C
Health Hazard 1Danger