2-chloroethylamine hydrochloride, 98%
2-chloroethylamine hydrochloride, 98%
2-chloroethylamine hydrochloride, 98%
2-chloroethylamine hydrochloride, 98%
Thermo Scientific Chemicals
2-chloroethylamine hydrochloride, 98%
CAS: 870-24-6 | C2H7Cl2N | 115.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA422861000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Chloroethylamine hydrochloride
CAS870-24-6
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0