Xylenes, 99%, ACS reagent
Xylenes, 99%, ACS reagent
Xylenes, 99%, ACS reagent
Xylenes, 99%, ACS reagent
Thermo Scientific Chemicals
Xylenes, 99%, ACS reagent
CAS: 1330-20-7 | C8H10
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
25 L
Catalog number FSA422685000
Price (MYR)
321.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
321.00
BOT
Add to cart
Specifications
Chemical Name or MaterialXylenes
Melting Point-34.0°C
CAS1330-20-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.