Trimyristin, 90%
Trimyristin, 90%
Trimyristin, 90%
Thermo Scientific Chemicals
Trimyristin, 90%
CAS: 555-45-3 | C45H86O6 | 722.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA422090050
Price (MYR)
654.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
654.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrimyristin
Name Note90%
CAS555-45-3
Melting Point54°C to 58°C
ColorWhite to Beige