2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
Thermo Scientific Chemicals
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
CAS: 6846-50-0 | C16H30O4 | 286.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number FSA422000010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2,2,4-Trimethyl-1,3-pentanediol diisobutyrate
Name Note98%
CAS6846-50-0
CAS Min %97.5
CAS Max %100.0