2,3,4-Trihydroxybenzaldehyde, 98%
2,3,4-Trihydroxybenzaldehyde, 98%
2,3,4-Trihydroxybenzaldehyde, 98%
2,3,4-Trihydroxybenzaldehyde, 98%
Thermo Scientific Chemicals
2,3,4-Trihydroxybenzaldehyde, 98%
CAS: 2144-08-3 | C7H6O4 | 154.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA421790250
Price (MYR)
1,314.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,314.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 3, 4-Trihydroxybenzaldehyde
CAS2144-08-3
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0