Tridodecylamine, 90%
Tridodecylamine, 90%
Tridodecylamine, 90%
Tridodecylamine, 90%
Thermo Scientific Chemicals
Tridodecylamine, 90%
CAS: 102-87-4 | C36H75N | 522 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA421621000
Price (MYR)
1,764.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,764.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTridodecylamine
Name Note95%
CAS102-87-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement