Trichloroethylene, 99.6%, ACS reagent, stabilized
Trichloroethylene, 99.6%, ACS reagent, stabilized
Trichloroethylene, 99.6%, ACS reagent, stabilized
Trichloroethylene, 99.6%, ACS reagent, stabilized
Trichloroethylene, 99.6%, ACS reagent, stabilized
Thermo Scientific Chemicals
Trichloroethylene, 99.6%, ACS reagent, stabilized
CAS: 79-01-6 | C2HCl3 | 131.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA421525000
Price (MYR)
461.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
461.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTrichloroethylene
CAS79-01-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause drowsiness or dizziness.
Harmful to aquatic life with long lasting effects.
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r
View more
Suitable for battery materials development.
RUO – Research Use Only