2,2,2-Tribromoethanol, 99%
2,2,2-Tribromoethanol, 99%
2,2,2-Tribromoethanol, 99%
2,2,2-Tribromoethanol, 99%
Thermo Scientific Chemicals
2,2,2-Tribromoethanol, 99%
CAS: 75-80-9 | C2H3Br3O | 282.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA421430500
Price (MYR)
1,147.00
BOT
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,147.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 2, 2-Tribromoethanol
CAS75-80-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0