Benzyl mercaptan, 99%
Benzyl mercaptan, 99%
Benzyl mercaptan, 99%
Benzyl mercaptan, 99%
Thermo Scientific Chemicals
Benzyl mercaptan, 99%
CAS: 100-53-8 | C7H8S | 124.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA421265000
Price (MYR)
2,036.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,036.00
BOT
Add to cart
Specifications
Chemical Name or MaterialBenzyl mercaptan
CAS100-53-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0