p-Toluenesulfonic acid, sodium salt
p-Toluenesulfonic acid, sodium salt
p-Toluenesulfonic acid, sodium salt
p-Toluenesulfonic acid, sodium salt
Thermo Scientific Chemicals
p-Toluenesulfonic acid, sodium salt
CAS: 657-84-1 | C7H7NaO3S | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA421235000
Price (MYR)
1,526.00
G
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,526.00
G
Add to cart
Specifications
Chemical Name or Materialp-Toluenesulfonic acid, sodium salt
CAS657-84-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
Health Hazard 3GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.