p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Catalog number FSA421215000
Price (MYR)
648.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
648.00
EA
Add to cart
Specifications
Chemical Name or Materialp-Toluenesulfonic acid monohydrate
CAS6192-52-5
Health Hazard 1Danger
Health Hazard 2GHS H Statement Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.