p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
p-Toluenesulfonic acid monohydrate, ACS reagent
Thermo Scientific Chemicals
p-Toluenesulfonic acid monohydrate, ACS reagent
CAS: 6192-52-5 | C7H10O4S | 190.213 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
Catalog number FSA421210250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or Materialp-Toluenesulfonic acid monohydrate
CAS6192-52-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for