o-Tolidine, 95%, pract
o-Tolidine, 95%, pract
o-Tolidine, 95%, pract
o-Tolidine, 95%, pract
Thermo Scientific Chemicals
o-Tolidine, 95%, pract
CAS: 119-93-7 | C14H16N2 | 212.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
250 g
Catalog number FSA421125000
Price (MYR)
3,417.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
3,417.00
BOT
Add to cart
Specifications
Chemical Name or Materialo-Tolidine
Name Note(pract), 95%
CAS119-93-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
May cause cancer.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
View more
Indicator
RUO – Research Use Only