DL-alpha-Tocopheryl acetate, 98%
DL-alpha-Tocopheryl acetate, 98%
DL-alpha-Tocopheryl acetate, 98%
DL-alpha-Tocopheryl acetate, 98%
Thermo Scientific Chemicals
DL-alpha-Tocopheryl acetate, 98%
CAS: 7695-91-2 | C31H52O3 | 472.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA421070050
Price (MYR)
625.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
625.00
EA
Add to cart
Specifications
Chemical Name or MaterialDL-α-Tocopheryl acetate
CAS7695-91-2
ColorGreen to Yellow
EINECS Number231-710-
Flash Point257°C