D-delta-Tocopherol, 93%
D-delta-Tocopherol, 93%
D-delta-Tocopherol, 93%
D-delta-Tocopherol, 93%
Thermo Scientific Chemicals
D-delta-Tocopherol, 93%
CAS: 119-13-1 | C27H46O2 | 402.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA421051000
Price (MYR)
1,050.00
BOT
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,050.00
BOT
Add to cart
Specifications
Chemical Name or MaterialD-δ-Tocopherol
CAS119-13-1
ColorBrown to Yellow
EINECS Number204-299-
Flash Point>93°C