D-alpha-Tocopherol, 97+%
D-alpha-Tocopherol, 97+%
D-alpha-Tocopherol, 97+%
D-alpha-Tocopherol, 97+%
Thermo Scientific Chemicals
D-alpha-Tocopherol, 97+%
CAS: 59-02-9 | C29H50O2 | 430.7 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA421031000100 mgGlass Bottle
Catalog number FSA421031000
Price (MYR)
714.00
EA
Add to cart
Quantity:
100 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
714.00
EA
Add to cart
Specifications
Chemical Name or MaterialD-α-Tocopherol
Name Note99%
CAS59-02-9
Health Hazard 1LIGHT SENSITIVE
Health Hazard 2AIR SENSITIVE