N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
Thermo Scientific Chemicals
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride, 98+%
CAS: 637-01-4 | C10H16N2·2HCl | 237.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA420601000
Price (MYR)
3,386.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,386.00
EA
Add to cart
Specifications
Chemical Name or MaterialN,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
CAS637-01-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0