N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
Thermo Scientific Chemicals
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine, 99%
CAS: 140-07-8 | C10H24N2O4 | 236.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA420481000
Price (MYR)
272.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
272.00
BOT
Add to cart
Specifications
Chemical Name or MaterialN,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine
CAS140-07-8
CAS Min %98.5
CAS Max %100.0
ColorColorless to Amber