Tetraethyl orthosilicate, 98%, AcroSeal™
Tetraethyl orthosilicate, 98%, AcroSeal™
Tetraethyl orthosilicate, 98%, AcroSeal™
Tetraethyl orthosilicate, 98%, AcroSeal™
Tetraethyl orthosilicate, 98%, AcroSeal™
Thermo Scientific Chemicals
Tetraethyl orthosilicate, 98%, AcroSeal™
CAS: 78-10-4 | C8H20O4Si | 208.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA420361000
Price (MYR)
217.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
217.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetraethyl orthosilicate
CAS78-10-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.