Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade
Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade
Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade
Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade
Thermo Scientific Chemicals
Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade
CAS: 5574-97-0 | C16H38NO4P | 339.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number FSA420171000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5574-97-0, 7732-18-5
Specifications
Boiling Point100.0°C
Chemical Name or MaterialTetrabutylammonium phosphate monobasic
CAS5574-97-0
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
CAS7732-18-5
View more
For ion pair chromatography, Phase transfer catalyst
RUO – Research Use Only