Tetrabromophenolphthalein ethyl ester, pure
Tetrabromophenolphthalein ethyl ester, pure
Tetrabromophenolphthalein ethyl ester, pure
Tetrabromophenolphthalein ethyl ester, pure
Thermo Scientific Chemicals
Tetrabromophenolphthalein ethyl ester, pure
CAS: 1176-74-5 | C22H14Br4O4 | 661.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA420070010
Price (MYR)
1,795.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,795.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTetrabromophenolphthalein ethyl ester
CAS1176-74-5
Melting Point208.0°C to 211.0°C
ColorRed to Yellow
EINECS Number214-645-2