Sucrose octaacetate, 98%
Sucrose octaacetate, 98%
Sucrose octaacetate, 98%
Sucrose octaacetate, 98%
Thermo Scientific Chemicals
Sucrose octaacetate, 98%
CAS: 126-14-7 | C28H38O19 | 678.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA419790010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSucrose octaacetate
CAS126-14-7
Melting Point>78.0°C
ColorCream-White to White
Density1.2800g/mL