D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
Thermo Scientific Chemicals
D(+)-Sucrose, 99+%, for biochemistry, DNAse, RNAse and protease free
D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose. | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number FSA419762500
Price (MYR)
250.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
250.00
EA
Add to cart
Specifications
Chemical Name or MaterialD(+)-Sucrose
Name NoteRNAse Free and DNAse Free
CAS57-50-1
Melting Point190.0°C to 192.0°C
ColorWhite
View more
General Description
• D(+)-Sucrose is a disaccharide carbohydrate
• It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose
Applications
• D(+)-Sucrose can be used for protein and nucleic acid purification
• Its fatty acid esters have laboratory research applications in transdermal systems
• It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
RUO – Research Use Only
General References:
  1. Jain, A.; Shore, A. M.; Jonnalagadda, S. C.; Ramanujachary, K. V.; Mugweru, A. Conversion of fructose, glucose, and sucrose to 5-hydroxymethyl-2-furfural over mesoporous zirconium phosphate catalyst. Appl. Catal. A 2015, 489, 72-76.
  2. Chen, Y.; Zhang, H.; Li, A.; Zhuo, K. Conductivities for ZnSO4/MgSO4+ sucrose/trehalose + water systems at 298.15 K. Fluid Phase Equilib. 2015, 388, 78-83.