Stoddard Solvent
Stoddard Solvent
Stoddard Solvent
Thermo Scientific Chemicals
Stoddard Solvent
CAS: 8052-41-3 | C8H15BrO2 | 223.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Catalog number FSA419730050
Price (MYR)
2,205.00
EA
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
2,205.00
EA
Add to cart
Specifications
Chemical Name or MaterialStoddard solvent
Melting Point-40.0°C
CAS8052-41-3
Health Hazard 1Danger
Health Hazard 2GHS P Statement May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.