Starch, extra pure, potato, powder
Starch, extra pure, potato, powder
Starch, extra pure, potato, powder
Starch, extra pure, potato, powder
Thermo Scientific Chemicals
Starch, extra pure, potato, powder
CAS: 9005-25-8 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA419695000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialStarch
ColorWhite
pH6 to 8 (50% aq. suspension)
CAS9005-25-8
Physical FormPowder