Sodium polyanetholesulfonate
Sodium polyanetholesulfonate
Sodium polyanetholesulfonate
Sodium polyanetholesulfonate
Thermo Scientific Chemicals
Sodium polyanetholesulfonate
CAS: 55963-78-5 | (C10H11NaO4S)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number FSA419631000
Price (MYR)
2,298.00
BOT
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
2,298.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium polyanetholesulfonate
CAS55963-78-5
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement:
Wear protective gloves/eye protection/face protection.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.