Sodium periodate, 99.8+%, ACS reagent
Sodium periodate, 99.8+%, ACS reagent
Sodium periodate, 99.8+%, ACS reagent
Sodium periodate, 99.8+%, ACS reagent
Thermo Scientific Chemicals
Sodium periodate, 99.8+%, ACS reagent
CAS: 7790-28-5 | INaO4 | 213.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA419611000
Price (MYR)
1,007.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,007.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium periodate
Melting Point300.0°C
CAS7790-28-5
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
Oxidation reagent
RUO – Research Use Only