Sodium methoxide, pure, titrant, (0.5 M in methanol)
Sodium methoxide, pure, titrant, (0.5 M in methanol)
Sodium methoxide, pure, titrant, (0.5 M in methanol)
Sodium methoxide, pure, titrant, (0.5 M in methanol)
Thermo Scientific Chemicals
Sodium methoxide, pure, titrant, (0.5 M in methanol)
CAS: 124-41-4 | CH3NaO | 54.024 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA419605000
Price (MYR)
754.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
754.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium methoxide
ColorColorless
Name Note0.5 M in methanol
CAS124-41-4
Boiling Point65.0°C