Diethyldithiocarbamic acid, silver salt, ACS reagent
Diethyldithiocarbamic acid, silver salt, ACS reagent
Diethyldithiocarbamic acid, silver salt, ACS reagent
Diethyldithiocarbamic acid, silver salt, ACS reagent
Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, ACS reagent
CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
10 g
Packaging:
Glass bottle
Catalog number FSA419330250
Price (MYR)
2,890.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,890.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDiethyldithiocarbamic acid, silver salt
CAS1470-61-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective glov