Silica gel, for chromatography, 0.030-0.200 mm, 60 A
Silica gel, for chromatography, 0.030-0.200 mm, 60 A
Silica gel, for chromatography, 0.030-0.200 mm, 60 A
Silica gel, for chromatography, 0.030-0.200 mm, 60 A
Thermo Scientific Chemicals
Silica gel, for chromatography, 0.030-0.200 mm, 60 A
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass Bottle
Catalog number FSA419292500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSilica gel
ColorWhite
Physical FormPowder
Name Note0.060-0.200mm, 60 A
CAS7631-86-9
View more
For column chromatography
RUO – Research Use Only