Selenium, 99+%, powder
Selenium, 99+%, powder
Selenium, 99+%, powder
Selenium, 99+%, powder
Thermo Scientific Chemicals
Selenium, 99+%, powder
CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA419275000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Boiling Point685°C
Chemical Name or MaterialSelenium
Melting Point217°C
Name Note<325 mesh
CAS7782-49-2