Rosin, gum
Rosin, gum
Rosin, gum
Thermo Scientific Chemicals
Rosin, gum
CAS: 8050-09-7 | C20H30O2 | 302.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA419175000
Price (MYR)
489.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
489.00
BOT
Add to cart
Specifications
Chemical Name or MaterialRosin, gum
Acidity163 to 173mg KOH/g
CAS8050-09-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause an allergic skin reaction.
View more
Adsorbent
RUO – Research Use Only